Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039598.1 | NP_001034687.1 | 4 - 5 | 383 | 78 | |
GenBank mRNA | DQ129668.1 | - | 4 - 5 | 383 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |