Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001296.5 | NP_001001296.1 | 2 - 3 | 298 | 60 | |
GenBank mRNA | BX272937.4 | - | 2 - 3 | 298 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |