Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045836.1 | NP_001039301.1 | 3 - 4 | 460 | 126 | |
XM_025148683.1 | XP_025004451.1 | 3 - 4 | 590 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Inventoried Probe spans exons |