Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048077.1 | NP_001041542.1 | 11 - 12 | 1425 | 69 | |
XM_015281391.2 | XP_015136877.1 | 9 - 10 | 1376 | 69 | ||
XM_015281392.2 | XP_015136878.1 | 13 - 14 | 3697 | 69 | ||
GenBank mRNA | AB243875.1 | - | 11 - 12 | 1425 | 69 | |
AB243876.1 | - | 9 - 10 | 1238 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |