Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083361.1 | NP_001076830.1 | 5 - 6 | 786 | 131 | |
XM_015275777.2 | XP_015131263.1 | 4 - 5 | 131 | |||
GenBank mRNA | EF379383.1 | - | 5 - 6 | 786 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |