Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097529.2 | NP_001090998.2 | 1 - 2 | 68 | 67 | |
XM_015298633.2 | XP_015154119.2 | 3 - 4 | 2260 | 67 | ||
XM_015298634.2 | XP_015154120.2 | 4 - 5 | 1828 | 67 | ||
XM_015298638.2 | XP_015154124.2 | 3 - 4 | 2246 | 67 | ||
GenBank mRNA | AM412311.1 | - | 1 - 2 | 68 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |