Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098607.1 | NP_001092077.1 | 3 - 4 | 2530 | 74 | |
XM_015278278.2 | XP_015133764.1 | 3 - 4 | 74 | |||
GenBank mRNA | EF175383.1 | - | 3 - 4 | 2530 | 74 | |
HM056233.1 | - | 3 - 4 | 513 | 74 | ||
HM056234.1 | - | 3 - 4 | 513 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |