Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101034.1 | NP_001094504.1 | 3 - 4 | 782 | 85 | |
GenBank mRNA | BX930481.1 | - | 2 - 3 | 239 | 85 | |
EF624351.1 | - | 3 - 4 | 782 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |