Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205148.4 | NP_990479.1 | 4 - 5 | 1733 | 80 | |
XM_015284820.2 | XP_015140306.1 | 5 - 6 | 1212 | 80 | ||
GenBank mRNA | S83278.2 | - | 4 - 5 | 1094 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |