Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190165.4 | NP_001177094.1 | 5 - 6 | 545 | 89 | |
NR_073182.3 | 5 - 6 | 534 | 89 | |||
XM_015289639.2 | XP_015145125.1 | 6 - 7 | 89 | |||
GenBank mRNA | DQ185931.1 | - | 5 - 6 | 89 | ||
DR429295.1 | - | 5 - 6 | 541 | 89 | ||
S78477.1 | - | 5 - 6 | 543 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |