Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMJD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114659.1 | NP_001108131.1 | 3 - 4 | 478 | 67 | |
GenBank mRNA | BX929829.2 | - | 2 - 3 | 416 | 67 | |
CR407278.1 | - | 2 - 3 | 274 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |