Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204265.1 | NP_989596.1 | 3 - 4 | 705 | 83 | |
XM_015294110.2 | XP_015149596.1 | 4 - 5 | 372 | 83 | ||
XM_015294111.2 | XP_015149597.1 | 4 - 5 | 334 | 83 | ||
XM_015294113.2 | XP_015149599.1 | 4 - 5 | 274 | 83 | ||
XM_015294114.2 | XP_015149600.1 | 4 - 5 | 339 | 83 | ||
GenBank mRNA | AB069964.1 | - | 3 - 4 | 705 | 83 | |
AF461016.1 | - | 4 - 5 | 267 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |