Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204310.2 | NP_989641.2 | 3 - 4 | 451 | 76 | |
XM_015284003.2 | XP_015139489.1 | 3 - 4 | 946 | 76 | ||
GenBank mRNA | U86764.1 | - | 3 - 4 | 451 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |