Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204338.1 | NP_989669.1 | 1 - 2 | 630 | 111 | |
GenBank mRNA | AF075441.1 | - | 1 - 2 | 129 | 111 | |
AF123389.1 | - | 1 - 2 | 630 | 111 | ||
KX688154.1 | - | 1 - 2 | 101 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |