Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204384.1 | NP_989715.1 | 8 - 9 | 923 | 92 | |
GenBank mRNA | AY298725.1 | - | 8 - 9 | 923 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |