Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204385.1 | NP_989716.1 | 3 - 4 | 308 | 69 | |
XM_015290232.2 | XP_015145718.1 | 3 - 4 | 293 | 69 | ||
XM_025152733.1 | XP_025008501.1 | 4 - 5 | 345 | 69 | ||
GenBank mRNA | AY297457.1 | - | 3 - 4 | 308 | 69 | |
BX929559.2 | - | 2 - 3 | 163 | 69 | ||
BX950639.2 | - | 3 - 4 | 308 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Inventoried Probe spans exons |