Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204418.1 | NP_989749.1 | 1 - 2 | 262 | 95 | |
GenBank mRNA | L08047.1 | - | 1 - 2 | 262 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |