Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204469.1 | NP_989800.1 | 4 - 5 | 654 | 104 | |
GenBank mRNA | AY339892.1 | - | 4 - 5 | 654 | 104 | |
BX950349.1 | - | 4 - 5 | 635 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |