Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204537.1 | NP_989868.2 | 2 - 3 | 642 | 91 | |
GenBank mRNA | AY128507.1 | - | 2 - 3 | 642 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |