Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205073.1 | NP_990404.1 | 2 - 3 | 162 | 90 | |
GenBank mRNA | X95074.1 | - | 162 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |