Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFI1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205091.1 | NP_990422.1 | 4 - 5 | 613 | 119 | |
GenBank mRNA | Y10898.1 | - | 613 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |