Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205267.1 | NP_990598.1 | 10 - 11 | 910 | 143 | |
GenBank mRNA | AH002463.2 | - | 470 | 143 | ||
CR732944.1 | - | 10 - 11 | 792 | 143 | ||
X17200.1 | - | 910 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |