Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006135.1 | NP_001006135.1 | 1 - 2 | 466 | 87 | |
GenBank mRNA | AJ719277.1 | - | 1 - 2 | 466 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |