Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006135.1 | NP_001006135.1 | 8 - 9 | 1214 | 83 | |
GenBank mRNA | AJ719277.1 | - | 8 - 9 | 1214 | 83 | |
CR391479.1 | - | 6 - 7 | 610 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |