Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006142.1 | NP_001006142.1 | 4 - 5 | 535 | 137 | |
XM_025154495.1 | XP_025010263.1 | 7 - 8 | 6288 | 137 | ||
GenBank mRNA | AJ719288.1 | - | 4 - 5 | 535 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |