Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006180.1 | NP_001006180.1 | 3 - 4 | 320 | 92 | |
GenBank mRNA | AJ720047.1 | - | 3 - 4 | 320 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |