Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1H21ORF59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006258.1 | NP_001006258.1 | 3 - 4 | 446 | 72 | |
GenBank mRNA | AJ720779.1 | - | 3 - 4 | 446 | 72 | |
CR523820.1 | - | 3 - 4 | 256 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |