Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006285.1 | NP_001006285.1 | 2 - 3 | 137 | 71 | |
XM_015296246.2 | XP_015151732.1 | 2 - 3 | 239 | 71 | ||
XM_025142189.1 | XP_024997957.1 | 2 - 3 | 425 | 71 | ||
GenBank mRNA | AJ720641.1 | - | 2 - 3 | 137 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |