Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006285.1 | NP_001006285.1 | 7 - 8 | 586 | 120 | |
XM_015296246.2 | XP_015151732.1 | 7 - 8 | 688 | 120 | ||
GenBank mRNA | AJ720641.1 | - | 7 - 8 | 586 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |