Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006570.1 | NP_001006570.1 | 2 - 3 | 456 | 117 | |
GenBank mRNA | AJ719805.1 | - | 2 - 3 | 456 | 117 | |
CR387916.1 | - | 2 - 3 | 342 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |