Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HACD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007828.1 | NP_001007829.1 | 7 - 8 | 712 | 137 | |
GenBank mRNA | AJ719527.1 | - | 7 - 8 | 712 | 137 | |
CR405973.1 | - | 2 - 3 | 203 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |