Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007884.1 | NP_001007885.1 | 1 - 2 | 205 | 142 | |
GenBank mRNA | AJ720189.1 | - | 1 - 2 | 205 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |