Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007884.1 | NP_001007885.1 | 4 - 5 | 490 | 63 | |
GenBank mRNA | AJ720189.1 | - | 4 - 5 | 490 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |