Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007970.1 | NP_001007971.1 | 3 - 4 | 300 | 111 | |
XM_015301808.2 | XP_015157294.1 | 3 - 4 | 438 | 111 | ||
GenBank mRNA | AJ719969.1 | - | 3 - 4 | 300 | 111 |
Gene Symbol: LOC112533280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025154317.1 | XP_025010085.1 | 2 - 3 | 258 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |