Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128062.1 | NP_001121534.1 | 2 - 3 | 769 | 67 | |
GenBank mRNA | EU647890.1 | - | 2 - 3 | 769 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Gallus gallus Probe spans exons |