Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130743.1 | NP_001124215.1 | 2 - 3 | 221 | 108 | |
XM_015285151.2 | XP_015140637.1 | 1 - 2 | 591 | 108 | ||
GenBank mRNA | AB368182.1 | - | 2 - 3 | 221 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Gallus gallus Probe spans exons |