Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159315.1 | NP_001152787.1 | 3 - 4 | 299 | 64 | |
XM_015284568.2 | XP_015140054.1 | 4 - 5 | 501 | 64 | ||
XM_015284574.2 | XP_015140060.1 | 5 - 6 | 702 | 64 | ||
XM_015284580.2 | XP_015140066.1 | 4 - 5 | 370 | 64 | ||
XM_025146984.1 | XP_025002752.1 | 4 - 5 | 479 | 64 | ||
XM_025146985.1 | XP_025002753.1 | 3 - 4 | 309 | 64 | ||
GenBank mRNA | CR389683.1 | - | 4 - 5 | 654 | 64 | |
CR390482.1 | - | 3 - 4 | 402 | 64 | ||
FM994924.1 | - | 3 - 4 | 299 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |