Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM20L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206977.1 | NP_996860.1 | 1 | 2346 | 98 | |
GenBank mRNA | AB088841.1 | - | 1 | 2346 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |