Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251128.1 | NP_001238057.1 | 6 - 7 | 1089 | 66 | |
GenBank mRNA | AF126550.1 | - | 6 - 7 | 1089 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Glycine max Probe spans exons |