Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250643.2 | NP_001237572.1 | 4 - 5 | 895 | 102 | |
GenBank mRNA | AF141602.1 | - | 4 - 5 | 900 | 102 | |
AK244280.1 | - | 4 - 5 | 918 | 102 | ||
AK286711.1 | - | 4 - 5 | 925 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Glycine max Probe spans exons |