Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250974.1 | NP_001237903.1 | 2 - 3 | 1631 | 98 | |
GenBank mRNA | AJ001091.1 | - | 2 - 3 | 1631 | 98 |
Gene Symbol: LOC100787735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK286404.1 | - | 2 - 3 | 1675 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |