Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249285.1 | NP_001236214.1 | 4 - 5 | 357 | 100 | |
GenBank mRNA | AJ276866.1 | - | 5 - 6 | 531 | 100 | |
AY230156.1 | - | 4 - 5 | 357 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Glycine max Probe spans exons |