Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ401230.1 | - | 2 - 3 | 319 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |