Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251255.1 | NP_001238184.1 | 6 - 7 | 477 | 89 | |
GenBank mRNA | L27418.1 | - | 6 - 7 | 477 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |