Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251385.1 | NP_001238314.1 | 8 - 9 | 855 | 95 | |
GenBank mRNA | BT096713.1 | - | 8 - 9 | 888 | 95 | |
BT098302.1 | - | 8 - 9 | 886 | 95 | ||
L38856.1 | - | 8 - 9 | 855 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |