Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249840.1 | NP_001236769.1 | 5 - 6 | 990 | 86 | |
GenBank mRNA | M64704.1 | - | 5 - 6 | 990 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |