Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U08469.1 | - | 9 - 10 | 1025 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |