Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251355.1 | NP_001238284.1 | 18 - 19 | 1310 | 83 | |
GenBank mRNA | AK246054.1 | - | 18 - 19 | 1265 | 83 | |
U12735.1 | - | 18 - 19 | 1310 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |