Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250768.1 | NP_001237697.1 | 9 - 10 | 1411 | 84 | |
GenBank mRNA | AM261476.1 | - | 9 - 10 | 1411 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Detect Genomic DNA Glycine max Probe spans exons |