Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250505.1 | NP_001237434.1 | 13 - 14 | 1374 | 67 | |
GenBank mRNA | DQ267699.1 | - | 13 - 14 | 1374 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Glycine max Probe spans exons |